Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ablim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103177.2 | NP_001096647.1 | 10 | 1240 | ||
NM_001103178.2 | NP_001096648.1 | 7 | 662 | |||
NM_001290813.1 | NP_001277742.1 | 10 | 1226 | |||
NM_001290815.1 | NP_001277744.1 | 10 | 1240 | |||
NM_001290816.1 | NP_001277745.1 | 10 | 1240 | |||
NM_178688.3 | NP_848803.3 | 12 | 1358 | |||
GenBank mRNA | AF404774.1 | - | 12 | 1358 | ||
AF404775.1 | - | 10 | 1000 | |||
AF404776.1 | - | 7 | 622 | |||
AK079331.1 | - | 6 | 480 | |||
AK122196.1 | - | 10 | 1240 | |||
AK143907.1 | - | 10 | 1010 | |||
AK171860.1 | - | 1137 | ||||
BC032216.1 | - | 1 | 35 | |||
BC141083.1 | - | 10 | 1220 | 9055695 |