Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178691.4 | NP_848806.2 | 2 | 878 | ||
GenBank mRNA | AK036938.1 | - | 2 | 879 | ||
AK036991.1 | - | 2 | 880 | |||
AK044678.1 | - | 2 | 674 | |||
AK053730.1 | - | 2 | 668 | |||
BC099948.1 | - | 2 | 671 | 30097649 | ||
BC139034.1 | - | 2 | 804 | 8862056 |