Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005027.3 | NP_005018.1 | 9, 10 | 1657 | ||
NR_073517.1 | 9, 10 | 1657 | ||||
GenBank mRNA | BC011917.2 | - | 1, 2 | 99 | ||
BC032647.1 | - | 8, 9 | 1385 | 5518348 | ||
BC070082.1 | - | 9, 10 | 1629 | 30343107 | ||
BC090249.1 | - | 9, 10 | 1589 | 6341766 | ||
X80907.1 | - | 8, 9 | 1358 |