Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipo9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153774.1 | NP_722469.1 | 23 | 3054 | ||
GenBank mRNA | AF273672.1 | - | 23 | 3002 | ||
AF273673.1 | - | 23 | 3002 | |||
AJ309238.1 | - | 23 | 2996 | |||
AK036687.1 | - | 23 | 3004 | |||
AK078784.1 | - | 1 | 13 | |||
AK173127.1 | - | 20 | 4943 | |||
BC056469.1 | - | 24 | 3202 | |||
BC079546.1 | - | 25 | 3169 | |||
BC098508.1 | - | 23 | 3004 | 30015513 |