Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039482.1 | NP_001034571.1 | 3 | 320 | ||
GenBank mRNA | AK053102.1 | - | 3 | 270 | ||
AK053371.1 | - | 3 | 284 | |||
AK054169.1 | - | 3 | 288 | |||
AK220547.1 | - | 3 | 299 | |||
BC019571.1 | - | 3 | 292 | 4235202 | ||
BC060183.1 | - | 3 | 249 |