Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tiprl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145513.4 | NP_663488.1 | 2 | 323 | ||
GenBank mRNA | AK049119.1 | - | 2 | 271 | ||
AK049784.1 | - | 2 | 309 | |||
AK075774.1 | - | 2 | 303 | |||
BC002098.1 | - | 2 | 268 | 3590157 | ||
BC058250.1 | - | 2 | 262 | 6315314 | ||
CF531065.1 | - | 2 | 325 |