Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tiprl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145513.4 | NP_663488.1 | 3 | 497 | ||
GenBank mRNA | AK049119.1 | - | 3 | 445 | ||
AK049784.1 | - | 3 | 483 | |||
AK075774.1 | - | 3 | 477 | |||
BC002098.1 | - | 3 | 442 | 3590157 | ||
BC058250.1 | - | 3 | 436 | 6315314 | ||
CF531065.1 | - | 3 | 499 |