Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005508.2 | NP_001005508.2 | 10 | 1398 | ||
GenBank mRNA | AK031319.1 | - | 10 | 1169 | ||
AK147757.1 | - | 10 | 1406 | |||
AK156351.1 | - | 10 | 1387 | |||
AK220204.1 | - | 5 | 532 | |||
BC082573.1 | - | 10 | 1346 | 30548035 |