Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146105.3 | NP_666217.2 | 3, 4 | 706 | ||
GenBank mRNA | AK036332.1 | - | 3, 4 | 738 | ||
AK087361.1 | - | 3, 4 | 724 | |||
AK146681.1 | - | 3, 4 | 796 | |||
BC039206.1 | - | 1, 2 | 69 | |||
BC054802.1 | - | 3, 4 | 706 | 6837803 |