Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mark1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145515.2 | NP_663490.2 | 13, 14 | 1546 | ||
GenBank mRNA | AB185282.1 | - | 13, 14 | 1469 | ||
AF453686.1 | - | 13, 14 | 1476 | |||
AK173181.1 | - | 12, 13 | 1396 | |||
BC112400.1 | - | 13, 14 | 1475 | 40059725 | ||
CX210969.1 | - | 5, 6 | 606 |