Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbspon | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033288.3 | NP_001028460.1 | 2 | 445 | ||
GenBank mRNA | AK137144.1 | - | 2 | 446 | ||
AK143251.1 | - | 2 | 419 | |||
BC147194.1 | - | 2 | 432 | 9056240 | ||
BC147195.1 | - | 2 | 432 | 9056224 |