Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PITPNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006224.3 | NP_006215.1 | 4, 6 | 545 | ||
XM_011523940.1 | XP_011522242.1 | 4, 6 | 967 | |||
GenBank mRNA | AK303986.1 | - | 4, 6 | 315 | ||
AL542612.3 | - | 4, 6 | 543 | |||
BC045108.1 | - | 4, 6 | 520 | 5454273 | ||
BC068538.1 | - | 4, 6 | 428 | |||
BC082976.1 | - | 4, 6 | 460 | 6578454 | ||
D30036.1 | - | 4, 6 | 497 | |||
EU794647.1 | - | 4, 6 | 520 | |||
M73704.1 | - | 4, 6 | 505 |