Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183019.2 | NP_898840.2 | 6 | 1113 | ||
GenBank mRNA | AK031310.1 | - | 6 | 1131 | ||
AK034013.1 | - | 6 | 1077 | |||
AK122448.2 | - | 2 | 168 | |||
AK134629.1 | - | 1076 | ||||
AK138598.1 | - | 6 | 1073 | |||
AK163424.1 | - | 6 | 1095 | |||
AK169449.1 | - | 5 | 1446 | |||
AK169957.1 | - | 6 | 1133 | |||
BC055804.1 | - | 6 | 1074 | |||
BC094608.1 | - | 4633 | ||||
BC139371.1 | - | 6 | 1096 | 8862393 | ||
BC139397.1 | - | 6 | 1096 | 8862419 |