Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PITPNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006224.3 | NP_006215.1 | 3 | 408 | ||
XM_011523939.1 | XP_011522241.1 | 3 | 395 | |||
XM_011523940.1 | XP_011522242.1 | 3 | 830 | |||
GenBank mRNA | AK303986.1 | - | 3 | 178 | ||
AL542612.3 | - | 3 | 406 | |||
BC027177.1 | - | 3 | 338 | |||
BC045108.1 | - | 3 | 383 | 5454273 | ||
BC068538.1 | - | 3 | 291 | |||
BC082976.1 | - | 3 | 323 | 6578454 | ||
D30036.1 | - | 3 | 360 | |||
EU794647.1 | - | 3 | 383 | |||
M73704.1 | - | 3 | 368 |