Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PITX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000325.5 | NP_000316.2 | 3 | 1181 | ||
NM_001204397.1 | NP_001191326.1 | 7 | 2345 | |||
NM_001204398.1 | NP_001191327.1 | 5 | 737 | |||
NM_001204399.1 | NP_001191328.1 | 4 | 599 | |||
NM_153426.2 | NP_700475.1 | 6 | 2111 | |||
NM_153427.2 | NP_700476.1 | 5 | 1973 | |||
XM_011532027.2 | XP_011530329.1 | 4 | 1849 | |||
GenBank mRNA | AF048720.1 | - | 4 | 634 | ||
AF048721.1 | - | 5 | 772 | |||
AF048722.1 | - | 3 | 1136 | |||
AK127829.1 | - | 7 | 2183 | |||
AK291591.1 | - | 4 | 627 | |||
AK313987.1 | - | 3 | 836 | |||
BC013998.2 | - | 3 | 1180 | 3937807 | ||
BC106010.1 | - | 4 | 609 | 3914727 | ||
U69961.1 | - | 5 | 967 |