Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PITX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005029.3 | NP_005020.1 | 3 | 326 | ||
GenBank mRNA | AF041339.1 | - | 2 | 175 | ||
BC011642.2 | - | 3 | 325 | 4131442 |