Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam134a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170755.2 | NP_739561.2 | 2 | 393 | ||
GenBank mRNA | AK034667.1 | - | 2 | 394 | ||
AK152268.1 | - | 2 | 108 | |||
AK158940.1 | - | 2 | 163 | |||
AK160400.1 | - | 2 | 347 | |||
AK165957.1 | - | 2 | 372 | |||
AK172093.1 | - | 2 | 390 | |||
BC070406.1 | - | 2 | 385 | 5707319 | ||
BY261213.1 | - | 2 | 171 |