Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033291.2 | NP_001028463.1 | 13 | 1795 | ||
NM_001198573.1 | NP_001185502.1 | 13 | 1795 | |||
GenBank mRNA | AK050239.1 | - | 13 | 1792 | ||
AK132295.1 | - | 13 | 1795 | |||
BC030843.1 | - | 13 | 1745 | |||
BC089376.1 | - | 10 | 1297 | |||
BC117957.1 | - | 14 | 1910 | 40098174 | ||
BC117958.1 | - | 13 | 1778 | 40098175 |