Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpp38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013376.2 | NP_001013394.2 | 2 | 365 | ||
GenBank mRNA | AK049133.1 | - | 2 | 310 | ||
AK087782.1 | - | 2 | 366 | |||
AK139023.1 | - | 2 | 293 | |||
BC051448.1 | - | 2 | 340 | 30060460 |