Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033294.3 | NP_001028466.2 | 19 | 2276 | ||
GenBank mRNA | AK030888.1 | - | 19 | 2223 | ||
AK144739.1 | - | 19 | 1990 | |||
AK166327.1 | - | 19 | 1946 | |||
BC066017.1 | - | 19 | 1913 | 6401415 | ||
BC132351.1 | - | 19 | 1966 | 40130628 |