Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206796.2 | NP_001193725.1 | 4 | 1059 | ||
NM_001206799.1 | NP_001193728.1 | 3 | 389 | |||
NM_001316318.1 | NP_001303247.1 | 3 | 942 | |||
NM_002654.5 | NP_002645.3 | 3 | 722 | |||
NM_182470.3 | NP_872270.1 | 3 | 938 | |||
NM_182471.3 | NP_872271.1 | 3 | 722 | |||
XM_005254443.1 | XP_005254500.1 | 4 | 446 | |||
XM_005254445.4 | XP_005254502.1 | 3 | 283 | |||
XM_006720570.1 | XP_006720633.1 | 4 | 543 | |||
XM_011521670.2 | XP_011519972.1 | 3 | 508 | |||
XM_017022313.1 | XP_016877802.1 | 3 | 283 | |||
GenBank mRNA | AK092369.1 | - | 3 | 472 | ||
AK222927.1 | - | 3 | 288 | |||
AK294315.1 | - | 4 | 548 | |||
AK299413.1 | - | 3 | 286 | |||
AK307855.1 | - | 4 | 289 | |||
AK309781.1 | - | 3 | 391 | |||
AK312253.1 | - | 3 | 288 | |||
BC000481.2 | - | 3 | 304 | 2964687 | ||
BC007640.1 | - | 3 | 288 | 3533876 | ||
BC007952.2 | - | 3 | 300 | 4299213 | ||
BC012811.2 | - | 3 | 300 | 2958817 | ||
BC023592.2 | - | 3 | 250 | |||
BC035198.1 | - | 3 | 254 | 3859987 | ||
BC096823.1 | - | 3 | 489 | |||
DA382029.1 | - | 3 | 529 | |||
DC326232.1 | - | 3 | 514 | |||
DC376054.1 | - | 3 | 484 | |||
EU794631.1 | - | 3 | 441 | |||
M23725.1 | - | 3 | 309 | |||
M26252.1 | - | 3 | 289 | |||
Z36831.1 | - | 3 | 327 |