Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290503.1 | NP_001277432.1 | 12 | 2180 | ||
NM_178694.4 | NP_848809.2 | 12 | 2219 | |||
GenBank mRNA | AK155401.1 | - | 12 | 2127 | ||
BC048924.1 | - | 12 | 2062 | 6312691 | ||
BC057021.1 | - | 12 | 2124 | 6825937 |