Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D2Wsu81e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172660.4 | NP_766248.3 | 11 | 1004 | ||
GenBank mRNA | AK036945.1 | - | 9 | 1298 | ||
AK044983.1 | - | 9 | 1598 | |||
AK046148.1 | - | 10 | 1199 | |||
AK147162.1 | - | 11 | 994 | |||
BC078441.1 | - | 8 | 1586 | 5686308 |