Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dolk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177648.3 | NP_808316.1 | 1 | 924 | ||
GenBank mRNA | AB073968.1 | - | 1 | 867 | ||
AK077581.1 | - | 1 | 894 | |||
AK144705.1 | - | 1 | 905 | |||
BC019543.1 | - | 1 | 869 | |||
BC026973.1 | - | 1 | 649 | 5371267 |