Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198304.2 | NP_938046.2 | 24 | 2476 | ||
GenBank mRNA | AK129073.1 | - | 24 | 2476 | ||
BC032090.1 | - | 24 | 2478 | |||
BC056489.1 | - | 10 | 1057 | |||
BC096591.1 | - | 24 | 2477 | 4916413 |