Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3glb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289709.1 | NP_001276638.1 | 3 | 331 | ||
NM_001289710.1 | NP_001276639.1 | 3 | 331 | |||
NM_139302.2 | NP_647463.1 | 3 | 331 | |||
GenBank mRNA | AF426314.1 | - | 3 | 228 | ||
AK129455.1 | - | 3 | 259 | |||
AK152409.1 | - | 3 | 261 | |||
AK155248.1 | - | 3 | 256 | |||
AK167762.1 | - | 3 | 304 | |||
AK182931.1 | - | 2 | 42 | |||
AK188040.1 | - | 2 | 41 | |||
BC024477.1 | - | 3 | 246 | 4977001 | ||
BC028859.1 | - | 3 | 320 | 4167974 | ||
BY317126.1 | - | 3 | 332 | |||
CX216029.1 | - | 3 | 321 | |||
EF676083.1 | - | 3 | 307 |