Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK083701.1 | - | 10 | 1350 | ||
BC034076.1 | - | 11 | 1780 | |||
BC040753.1 | - | 11 | 1780 |