Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qrfp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183424.3 | NP_906269.1 | 2 | 767 | ||
GenBank mRNA | AK029144.1 | - | 2 | 768 |