Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKNOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286258.1 | NP_001273187.1 | 4 | 440 | ||
NM_001320694.1 | NP_001307623.1 | 5 | 612 | |||
NM_004571.4 | NP_004562.2 | 5 | 612 | |||
GenBank mRNA | AK222892.1 | - | 4 | 478 | ||
AK294794.1 | - | 4 | 309 | |||
AY142115.1 | - | 5 | 589 | |||
AY196965.1 | - | 5 | 590 | |||
AY566249.1 | - | 4 | 460 | |||
BC000735.2 | - | 5 | 558 | |||
BC007746.2 | - | 4 | 419 | 4099156 | ||
DR000771.1 | - | 5 | 539 | |||
U68727.1 | - | 4 | 483 | |||
Y13613.1 | - | 5 | 525 |