Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam129b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146119.2 | NP_666231.1 | 10 | 1498 | ||
GenBank mRNA | AK033735.1 | - | 10 | 1466 | ||
AK046583.1 | - | 10 | 1395 | |||
AK051396.1 | - | 10 | 1437 | |||
AK146953.1 | - | 10 | 1483 | |||
AK149818.1 | - | 10 | 1409 | |||
AK149888.1 | - | 10 | 1408 | |||
AK150642.1 | - | 10 | 1436 | |||
AK152207.1 | - | 10 | 1457 | |||
AK153489.1 | - | 10 | 1442 | |||
AK170512.1 | - | 10 | 1438 | |||
AK196560.1 | - | 1 | 17 | |||
BC024639.1 | - | 2 | 231 | |||
BC027843.1 | - | 10 | 1418 | 5343013 |