Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam129b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146119.2 | NP_666231.1 | 5 | 723 | ||
GenBank mRNA | AK033735.1 | - | 5 | 691 | ||
AK046583.1 | - | 5 | 620 | |||
AK051396.1 | - | 5 | 662 | |||
AK146953.1 | - | 5 | 708 | |||
AK149818.1 | - | 5 | 634 | |||
AK149888.1 | - | 5 | 633 | |||
AK150642.1 | - | 5 | 661 | |||
AK152207.1 | - | 5 | 682 | |||
AK153489.1 | - | 5 | 667 | |||
AK170512.1 | - | 5 | 663 | |||
AK177466.1 | - | 2 | 168 | |||
AK206543.1 | - | 2 | 109 | |||
AK214664.1 | - | 2 | 109 | |||
AK217216.1 | - | 2 | 109 | |||
BC027843.1 | - | 5 | 643 | 5343013 |