Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapkap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290625.1 | NP_001277554.1 | 11 | 1507 | ||
NM_001290626.1 | NP_001277555.1 | 10 | 1505 | |||
NM_177345.4 | NP_796319.1 | 11 | 1833 | |||
GenBank mRNA | AK027932.1 | - | 6 | 606 | ||
AK042154.1 | - | 10 | 1495 | |||
AK052045.1 | - | 11 | 1501 | |||
AK132263.1 | - | 11 | 1497 | |||
BC027377.1 | - | 6 | 635 | |||
BC031579.1 | - | 6 | 635 | |||
BC043296.1 | - | 11 | 1732 | |||
BC090644.1 | - | 10 | 1407 | 30629263 | ||
BC096618.1 | - | 11 | 1806 | 6392317 |