Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000668.5 | NP_000659.2 | 8 | 1159 | ||
NM_001286650.1 | NP_001273579.1 | 9 | 1281 | |||
GenBank mRNA | AF153821.1 | - | 8 | 1116 | ||
AK299409.1 | - | 9 | 1077 | |||
AK301018.1 | - | 10 | 1219 | |||
AK303945.1 | - | 7 | 949 | |||
AK304396.1 | - | 8 | 883 | |||
AK316501.1 | - | 8 | 949 | |||
BC033009.2 | - | 8 | 1118 | 4828235 | ||
BX647469.1 | - | 8 | 3143 | |||
D00137.1 | - | 8 | 1068 | |||
FJ224285.1 | - | 8 | 1118 | |||
M21692.1 | - | 8 | 1080 | |||
M24317.1 | - | 8 | 1078 | |||
X03350.1 | - | 8 | 1120 |
Gene Symbol: ADH1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000667.3 | NP_000658.1 | 8 | 1162 | ||
GenBank mRNA | AK290558.1 | - | 8 | 1119 | ||
BC074738.2 | - | 8 | 1076 | 30915195 | ||
BC117442.1 | - | 8 | 1118 | 40125993 | ||
BC126306.1 | - | 8 | 1118 | 8992022 | ||
BG199182.1 | - | 2 | 225 | |||
BT019812.1 | - | 8 | 1048 | |||
M12271.1 | - | 8 | 1063 | |||
M12963.1 | - | 8 | 1120 |