Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206367.1 | NP_001193296.1 | 11 | 1484 | ||
NM_001206368.1 | NP_001193297.1 | 10 | 1400 | |||
NM_001206369.1 | NP_001193298.1 | 10 | 1445 | |||
NM_146120.4 | NP_666232.2 | 10 | 1507 | |||
GenBank mRNA | AK076156.1 | - | 10 | 1447 | ||
AK089934.1 | - | 11 | 1439 | |||
AK135632.1 | - | 9 | 1220 | |||
AK143664.1 | - | 10 | 1474 | |||
AK146969.1 | - | 11 | 1401 | |||
AK148957.1 | - | 11 | 1464 | |||
AK150359.1 | - | 11 | 1407 | |||
AK151684.1 | - | 11 | 1426 | |||
AK153029.1 | - | 11 | 1404 | |||
AK166831.1 | - | 10 | 1420 | |||
AK166932.1 | - | 11 | 1398 | |||
AK167918.1 | - | 11 | 1414 | |||
AK168461.1 | - | 11 | 1417 | |||
AK168532.1 | - | 11 | 1418 | |||
AK168707.1 | - | 10 | 1353 | |||
AK168725.1 | - | 11 | 1401 | |||
AK168829.1 | - | 10 | 1452 | |||
AK169041.1 | - | 11 | 1404 | |||
AK169670.1 | - | 11 | 1429 | |||
BC023143.1 | - | 10 | 1445 | 3711602 | ||
BC060377.1 | - | 11 | 1432 | 4983664 | ||
J04953.1 | - | 9 | 1294 |