Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172663.4 | NP_766251.3 | 9, 10 | 1708 | ||
GenBank mRNA | AK031025.1 | - | 9, 10 | 1681 | ||
AK047947.1 | - | 9, 10 | 1376 | |||
AK141073.1 | - | 999 | ||||
AK164886.1 | - | 5, 6 | 735 | |||
BC048785.1 | - | 8, 9 | 1147 |