Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081087.1 | NP_001074556.1 | 1 | 373 | ||
GenBank mRNA | AK132646.1 | - | 1 | 374 | ||
BC150698.1 | - | 1 | 458 | 9087609 |