Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cerkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048176.1 | NP_001041641.1 | 2 | 438 | ||
GenBank mRNA | BC046474.1 | - | 2 | 544 | ||
BY742285.1 | - | 2 | 448 | |||
HQ426680.1 | - | 2 | 438 | |||
HQ426681.1 | - | 2 | 438 | |||
HQ426682.1 | - | 2 | 438 | |||
HQ426684.1 | - | 2 | 438 | |||
HQ426685.1 | - | 2 | 438 | |||
HQ426686.1 | - | 2 | 438 | |||
HQ426687.1 | - | 2 | 569 | |||
HQ426688.1 | - | 2 | 438 | |||
HQ426689.1 | - | 2 | 438 | |||
HQ426690.1 | - | 2 | 438 | |||
HQ426691.1 | - | 237 | ||||
HQ426692.1 | - | 368 | ||||
HQ426693.1 | - | 278 | ||||
HQ426694.1 | - | 237 | ||||
HQ426695.1 | - | 410 | ||||
HQ426696.1 | - | 410 | ||||
HQ426697.1 | - | 410 | ||||
HQ426698.1 | - | 410 | ||||
HQ426699.1 | - | 410 | ||||
HQ426700.1 | - | 541 | ||||
HQ426701.1 | - | 410 |