Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172673.3 | NP_766261.2 | 13 | 1281 | ||
GenBank mRNA | AK030883.1 | - | 13 | 1037 | ||
AK044270.1 | - | 13 | 1032 | |||
AK146383.1 | - | 12 | 1161 | |||
BC038623.1 | - | 13 | 1018 | |||
BC057549.1 | - | 10 | 810 | 6416914 | ||
BC062889.1 | - | 13 | 1238 | 5698263 |