Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001311193.1 | NP_001298122.1 | 13 | 1702 | ||
NM_024420.2 | NP_077734.1 | 15 | 1882 | |||
XM_005245267.3 | XP_005245324.2 | 13 | 1703 | |||
XM_011509642.2 | XP_011507944.1 | 15 | 1833 | |||
GenBank mRNA | AK290336.1 | - | 15 | 1833 | ||
AK302938.1 | - | 13 | 1612 | |||
BC114340.2 | - | 15 | 1779 | 40034995 | ||
M68874.1 | - | 15 | 1815 | |||
M72393.1 | - | 15 | 1802 |