Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptlc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175467.3 | NP_780676.1 | 13 | 1830 | ||
GenBank mRNA | AK048374.1 | - | 12 | 1691 | ||
AK054240.1 | - | 12 | 1917 | |||
AK078679.1 | - | 12 | 1894 | |||
AK078686.1 | - | 13 | 1831 | |||
BC094496.1 | - | 12 | 1675 | 3483569 |