Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csrp2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166640.1 | NP_001160112.1 | 8 | 1817 | ||
NM_181417.3 | NP_852082.2 | 8 | 2300 | |||
GenBank mRNA | AK191012.1 | - | 1 | 82 | ||
BC031563.1 | - | 8 | 2300 | 4949236 | ||
BC033464.1 | - | 8 | 1817 |