Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031363.1 | - | 4 | 577 | ||
AK032644.1 | - | 3 | 465 | |||
AK045891.1 | - | 4 | 560 | |||
AK053924.1 | - | 4 | 578 | |||
AK078035.1 | - | 2 | 325 | |||
AK078867.1 | - | 4 | 536 | |||
AK083431.1 | - | 2 | 341 | |||
BC012260.1 | - | 4 | 539 | |||
BC128314.1 | - | 3 | 322 | |||
BC131928.1 | - | 4 | 441 | 40130205 | ||
BC131930.1 | - | 4 | 441 | 40130207 |