Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031363.1 | - | 6 | 810 | ||
AK045891.1 | - | 6 | 793 | |||
AK050529.1 | - | 1 | 495 | |||
AK053924.1 | - | 6 | 811 | |||
AK078035.1 | - | 4 | 563 | |||
AK078867.1 | - | 6 | 769 | |||
AK083431.1 | - | 4 | 574 | |||
BC012260.1 | - | 6 | 770 | |||
BC128314.1 | - | 5 | 555 | |||
BC131928.1 | - | 6 | 674 | 40130205 | ||
BC131930.1 | - | 6 | 674 | 40130207 |