Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172674.2 | NP_766262.2 | 5, 6 | 611 | ||
GenBank mRNA | AK033017.1 | - | 4, 5 | 410 | ||
AK038573.1 | - | 5, 6 | 518 | |||
AK045309.1 | - | 5, 6 | 530 | |||
BC060121.1 | - | 5, 6 | 491 | 6413021 |