Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146129.3 | NP_666241.1 | 9 | 1174 | ||
GenBank mRNA | AK089358.1 | - | 9 | 1175 | ||
AK150740.1 | - | 8 | 1065 | |||
BC031431.1 | - | 9 | 1143 | 4241283 |