Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000930.4 | NP_000921.1 | 10 | 1183 | ||
NM_001319189.1 | NP_001306118.1 | 8 | 916 | |||
NM_033011.3 | NP_127509.1 | 9 | 1045 | |||
GenBank mRNA | A03776.1 | - | 9 | 960 | ||
A07197.1 | - | 9 | 950 | |||
AF260825.1 | - | 4 | 425 | |||
AK289387.1 | - | 10 | 1135 | |||
AK290575.1 | - | 10 | 1135 | |||
AK297738.1 | - | 8 | 757 | |||
AK297937.1 | - | 7 | 646 | |||
AK299205.1 | - | 10 | 1025 | |||
AK300984.1 | - | 10 | 835 | |||
AK313342.1 | - | 9 | 886 | |||
AY221101.1 | - | 9 | 926 | |||
BC002795.2 | - | 9 | 871 | 3618149 | ||
BC007231.1 | - | 10 | 1007 | 3161005 | ||
BC013968.2 | - | 10 | 1009 | 3163742 | ||
BC018636.2 | - | 10 | 1009 | 3637273 | ||
BC095403.1 | - | 10 | 1024 | 30528441 | ||
BT007060.1 | - | 9 | 926 | |||
D01096.1 | - | 8 | 838 | |||
FM956488.1 | - | 9 | 937 | |||
M15518.1 | - | 10 | 1002 | |||
M18182.1 | - | 9 | 939 | |||
V00570.1 | - | 3 | 182 | |||
X02901.1 | - | 9 | 943 | |||
X07393.1 | - | 10 | 979 |