Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbln4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175631.3 | NP_783439.1 | 2 | 1464 | ||
GenBank mRNA | AK032406.1 | - | 2 | 968 | ||
AK032621.1 | - | 2 | 1467 | |||
AY134663.1 | - | 2 | 296 | |||
BC094540.1 | - | 2 | 1367 | 5708067 | ||
BC132025.1 | - | 2 | 562 | 40130302 | ||
BC132027.1 | - | 2 | 562 | 40130304 |