Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000930.4 | NP_000921.1 | 6, 7 | 801 | ||
NM_033011.3 | NP_127509.1 | 5, 6 | 663 | |||
GenBank mRNA | A03776.1 | - | 5, 6 | 578 | ||
A07197.1 | - | 5, 6 | 568 | |||
AK289387.1 | - | 6, 7 | 753 | |||
AK290575.1 | - | 6, 7 | 753 | |||
AK299205.1 | - | 6, 7 | 643 | |||
AK313342.1 | - | 5, 6 | 504 | |||
AY221101.1 | - | 5, 6 | 544 | |||
BC002795.2 | - | 5, 6 | 489 | 3618149 | ||
BC007231.1 | - | 6, 7 | 625 | 3161005 | ||
BC013968.2 | - | 6, 7 | 627 | 3163742 | ||
BC018636.2 | - | 6, 7 | 627 | 3637273 | ||
BC095403.1 | - | 6, 7 | 642 | 30528441 | ||
BT007060.1 | - | 5, 6 | 544 | |||
D01096.1 | - | 4, 5 | 456 | |||
FM956488.1 | - | 5, 6 | 555 | |||
M15518.1 | - | 6, 7 | 620 | |||
M18182.1 | - | 5, 6 | 557 | |||
X02901.1 | - | 5, 6 | 561 | |||
X07393.1 | - | 6, 7 | 597 | |||
X13097.1 | - | 6, 7 | 652 |