Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144500.3 | NP_653083.2 | 10 | 1143 | ||
GenBank mRNA | AK048537.1 | - | 10 | 1063 | ||
AK145292.1 | - | 10 | 1175 | |||
AK161695.1 | - | 10 | 1082 | |||
AK166059.1 | - | 10 | 1122 | |||
AK170306.1 | - | 10 | 1127 | |||
AK170669.1 | - | 10 | 1144 | |||
AK178638.1 | - | 4 | 322 | |||
AK182769.1 | - | 4 | 331 | |||
AK220257.1 | - | 4 | 462 | |||
BC026804.1 | - | 10 | 1150 | 4159217 |