Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmeb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198169.2 | NP_937812.1 | 9 | 979 | ||
GenBank mRNA | AB221583.1 | - | 8 | 860 | ||
AB221584.1 | - | 8 | 867 | |||
AK129319.1 | - | 5 | 511 | |||
BC029763.1 | - | 8 | 802 | |||
BC059066.1 | - | 9 | 979 | 6825428 |